Dimethyl 2,5-dihydroxyterephthalate 98%
Supplier: Ambeed
|
Warning
|
A208067-1G
A208067-5G
77112-356EA
12.74
USD
77112-356
77112-358
Dimethyl 2,5-dihydroxyterephthalate 98%
Dimethyl 2,5-dihydroxyterephthalate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₆ MW: 226.18 g/mol |
MDL Number:
MFCD00157193 CAS Number: 5870-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...