Triclabendazole 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)-1H-benzo[d]imidazole, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole, 6-Chloro-5-(2,3-dichlorophenoxy)-2-(methylthio)benzimidazole

A206752-10G A206752-100G A206752-25G
77112-342EA 21.22 USD
77112-342 77612-026 77612-028
Triclabendazole 98%
Triclabendazole

Precaución: for Research Use Only

Formula: C₁₄H₉Cl₃N₂OS
MW: 359.66 g/mol
MDL Number: MFCD00864519
CAS Number: 68786-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR