Sodium (S)-2-(4-(2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioate 99.9%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pemetrexed disodium
A206176-100MG
A206176-10MG
A206176-250MG
A206176-50MG
A206176-1MG
77112-324EA
12.74
USD
77112-324
77112-326
77112-328
77112-330
77685-531
Sodium (S)-2-(4-(2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioate 99.9%
Sodium (S)-2-(4-(2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioate
Precaución: for Research Use Only
Formula:
C₂₀H₁₉N₅Na₂O₆ MW: 471.37 g/mol |
MDL Number:
MFCD07779402 CAS Number: 150399-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...