5-Amino-4-methylpyrimidine 97%

Supplier: Ambeed
Warning

A203742-10G A203742-1G A203742-250MG A203742-5G
77112-264EA 295.61 USD
77112-264 77112-266 77112-268 77112-270
5-Amino-4-methylpyrimidine 97%
5-Amino-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MDL Number: MFCD09038780
CAS Number: 3438-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR