3-Methylpicolinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methylpyridine-2-carboxylic acid

A202118-100G A202118-10G A202118-25G A202118-500G A202118-5G
77112-190EA 81.69 USD
77112-190 77112-192 77112-194 77112-196 77112-198
3-Methylpicolinic acid 97%
3-Methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00191553
CAS Number: 4021-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR