2-Picolyl chloride hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Chloromethyl)pyridine HCl, 2-(Chloromethyl)pyridine hydrochloride

A202106-10G A202106-25G A202106-100G
77112-186EA 20.6 USD
77112-186 77112-188 77593-312
2-Picolyl chloride hydrochloride 98%
2-Picolyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₂N
MDL Number: MFCD00012811
CAS Number: 6959-47-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR