2-(Trifluoromethoxy)phenylacetylene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Ethynylphenyl trifluoromethyl ether, 1-Ethynyl-2-(trifluoromethoxy)benzene

A202092-1G A202092-250MG
77112-182EA 66.84 USD
77112-182 77112-184
2-(Trifluoromethoxy)phenylacetylene 95%
2-(Trifluoromethoxy)phenylacetylene

Precaución: for Research Use Only

Formula: C₉H₅F₃O
MW: 186.131 g/mol
MDL Number: MFCD03839994
CAS Number: 886363-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR