4-Bromo-3-fluoro-2-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-6-carboxy-2-fluorophenol, 4-Bromo-3-fluorosalicylic acid

A201991-100MG A201991-250MG
77112-178EA 68.96 USD
77112-178 77112-180
4-Bromo-3-fluoro-2-hydroxybenzoic acid 95%
4-Bromo-3-fluoro-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrFO₃
MW: 235.007 g/mol
MDL Number: MFCD23703631
CAS Number: 1429321-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR