2-Aminoacetonitrile hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: Aminoacetonitrile hydrochloride

A201541-100G A201541-25G
77112-176EA 41.17 USD
77112-176 77302-764
2-Aminoacetonitrile hydrochloride 98%
2-Aminoacetonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₂H₅ClN₂
MW: 92.53 g/mol
MDL Number: MFCD00012850
CAS Number: 6011-14-9
UN: 3439
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR