(S)-(+)-2-Amino-1-propanol 98%

Supplier: Ambeed
Danger

Synonyms: L(+)-Alaninol, L-Alaninol, (S)-2-Aminopropan-1-ol, L(+)-2-Amino-1-propanol

A201243-10G A201243-25G A201243-100G
77112-170EA 7.21 USD
77112-170 77112-172 77112-174 77593-304
(S)-(+)-2-Amino-1-propanol 98%
(S)-(+)-2-Amino-1-propanol

Precaución: for Research Use Only

Formula: C₃H₉NO
MDL Number: MFCD00064412
CAS Number: 2749-11-3
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR