6-Chloro-2-methyl-3-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-methylpyridine-3-boronic acid

A200010-1G A200010-250MG A200010-25G A200010-5G
77112-138EA 33.95 USD
77112-138 77112-140 77112-142 77112-144
6-Chloro-2-methyl-3-pyridineboronic acid 97%
6-Chloro-2-methyl-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₂
MW: 171.389 g/mol
MDL Number: MFCD07368875
CAS Number: 913836-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR