Di(N-succinimidyl) carbonate 95%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate

A199084-100G A199084-10G A199084-1G A199084-25G A199084-500G A199084-5G
77112-092EA 26.4 USD
77112-092 77112-094 77112-096 77112-098 77112-100 77112-102
Di(N-succinimidyl) carbonate 95%
Di(N-succinimidyl) carbonate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₇
MW: 256.17 g/mol
MDL Number: MFCD00009767
CAS Number: 74124-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR