4-Bromo-o-cresol 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-methylphenol, 1-Bromo-4-hydroxy-3-methylbenzene

A198010-100G A198010-10G A198010-1G A198010-25G A198010-5G A198010-500G
77112-056EA 88.06 USD
77112-056 77112-058 77112-060 77112-062 77112-064 77611-898
4-Bromo-o-cresol 95%
4-Bromo-o-cresol

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.04 g/mol
MDL Number: MFCD00055435
CAS Number: 2362-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR