(R)-2,3-Diaminopropanoic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-D-alanine Hydrochloride, H-D-Dap-OH.HCl, H-D-Dap-OH·HCl

A197656-10G A197656-1G A197656-250MG A197656-25G A197656-5G
77112-046EA 75.33 USD
77112-046 77112-048 77112-050 77112-052 77112-054
(R)-2,3-Diaminopropanoic acid hydrochloride 95%
(R)-2,3-Diaminopropanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₃H₉ClN₂O₂
MW: 140.5688 g/mol
MDL Number: MFCD00070606
CAS Number: 6018-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR