Triethyl phosphonoacetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate

A197335-1000G A197335-100G A197335-25G A197335-500G
77112-018EA 36.05 USD
77112-018 77112-020 77112-022 77112-024 77112-026
Triethyl phosphonoacetate 98%
Triethyl phosphonoacetate

Precaución: for Research Use Only

Formula: C₈H₁₇O₅P
MDL Number: MFCD00009177
CAS Number: 867-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR