1-(Difluoromethyl)-4-iodobenzene 95%

Supplier: Ambeed
Warning

A196945-100MG A196945-1G A196945-250MG A196945-5G
77112-008EA 55.17 USD
77112-008 77112-010 77112-012 77112-014
1-(Difluoromethyl)-4-iodobenzene 95%
1-(Difluoromethyl)-4-iodobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂I
MW: 254.02 g/mol
MDL Number: MFCD14698605
CAS Number: 1214372-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR