2-Bromo-3-fluoro-4-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-fluoro-4-picoline

A195989-100G A195989-10G A195989-1G A195989-250MG A195989-25G A195989-5G
77111-990EA 524.27 USD
77111-990 77111-992 77111-994 77111-996 77111-998 77112-000
2-Bromo-3-fluoro-4-methylpyridine 97%
2-Bromo-3-fluoro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01
MDL Number: MFCD04112499
CAS Number: 884494-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR