Ethyl 2-(2-phenyl-1H-indol-1-yl)acetate 95%

Supplier: Ambeed
Warning

A195964-100MG A195964-1G A195964-250MG A195964-5G
77111-982EA 33.95 USD
77111-982 77111-984 77111-986 77111-988
Ethyl 2-(2-phenyl-1H-indol-1-yl)acetate 95%
Ethyl 2-(2-phenyl-1H-indol-1-yl)acetate

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₂
MW: 279.33 g/mol
MDL Number: MFCD11046346
CAS Number: 65746-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR