(2-Bromoethoxy)benzene 98%

Supplier: Ambeed
Warning

Synonyms: β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole

A195744-100G A195744-500G A195744-10G A195744-25G
77111-978EA 52.53 USD
77111-978 77111-980 77139-230 77139-232
(2-Bromoethoxy)benzene 98%
(2-Bromoethoxy)benzene

Precaución: for Research Use Only

Formula: BrCH₂CH₂OC₆H₅
MW: 201.06 g/mol
Boiling Pt: 144
Flash Pt: 65
MDL Number: MFCD00000234
CAS Number: 589-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR