4-Amino-2-methyl-3-nitropyridine 97%

Supplier: Ambeed
Warning

A195538-100MG A195538-1G A195538-250MG
77111-950EA 13.8 USD
77111-950 77111-952 77111-954
4-Amino-2-methyl-3-nitropyridine 97%
4-Amino-2-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.1400 g/mol
MDL Number: MFCD08277309
CAS Number: 27582-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR