Methyl 3-(4-formylphenyl)acrylate 95%

Supplier: Ambeed
Warning

A194039-100G A194039-1G A194039-25G
77111-902EA 217.33 USD
77111-902 77111-904 77111-906
Methyl 3-(4-formylphenyl)acrylate 95%
Methyl 3-(4-formylphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₃
MDL Number: MFCD00157179
CAS Number: 58045-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR