N(α)-Benzyloxycarbonyl-D-lysine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-D-Lys-OH, Z-D-Lys-OH
A193482-10G
A193482-1G
A193482-25G
77111-870EA
81.37
USD
77111-870
77111-872
77111-874
N(α)-Benzyloxycarbonyl-D-lysine 95%
N(α)-Benzyloxycarbonyl-D-lysine
Precaución: for Research Use Only
Formula:
C₁₄H₂₀N₂O₄ MW: 280.3 g/mol |
MDL Number:
MFCD00067713 CAS Number: 70671-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...