6-Bromo-4-iodoquinoline 97%

Supplier: Ambeed
Warning

A193372-10G A193372-1G A193372-250MG A193372-5G
77111-862EA 755.37 USD
77111-862 77111-864 77111-866 77111-868
6-Bromo-4-iodoquinoline 97%
6-Bromo-4-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrIN
MW: 333.9512 g/mol
MDL Number: MFCD08437008
CAS Number: 927801-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR