2-Hydroxy-N-(4-hydroxyphenyl)benzamide 98%

Supplier: Ambeed
Warning

A193150-100G A193150-10G A193150-1G A193150-25G A193150-25MG A193150-500G A193150-5G
77111-848EA 211.12 USD
77111-848 77111-850 77111-852 77111-854 77111-856 77111-858 77111-860
2-Hydroxy-N-(4-hydroxyphenyl)benzamide 98%
2-Hydroxy-N-(4-hydroxyphenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₃
MW: 229.23 g/mol
MDL Number: MFCD00020026
CAS Number: 526-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR