3-Chloro-2,6-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A192520-100G A192520-10G A192520-1G A192520-250MG A192520-25G A192520-5G
77111-824EA 401.03 USD
77111-824 77111-826 77111-828 77111-830 77111-832 77111-834
3-Chloro-2,6-difluorobenzaldehyde 98%
3-Chloro-2,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
MDL Number: MFCD01631323
CAS Number: 190011-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR