1-Chloro-6,6-dimethyl-2-hepten-4-yne 95%

Supplier: Ambeed
Warning

A192431-100G A192431-1G A192431-500G A192431-5G
77111-812EA 49.44 USD
77111-812 77111-814 77111-816 77111-818
1-Chloro-6,6-dimethyl-2-hepten-4-yne 95%
1-Chloro-6,6-dimethyl-2-hepten-4-yne

Precaución: for Research Use Only

Formula: C₉H₁₃Cl
MDL Number: MFCD04039160
CAS Number: 126764-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR