3-Chloro-4-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chlor-3-cyan-6-fluorbenzene

A191541-100G A191541-10G A191541-25G A191541-5G
77111-778EA 61.54 USD
77111-778 77111-780 77111-782 77111-784
3-Chloro-4-fluorobenzonitrile 97%
3-Chloro-4-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClFN
MW: 155.56 g/mol
MDL Number: MFCD00015431
CAS Number: 117482-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR