N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid 96%

Supplier: Ambeed
Warning

A191498-100G A191498-10G A191498-1G A191498-25G A191498-5G
77111-768EA 227.63 USD
77111-768 77111-770 77111-772 77111-774 77111-776
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid 96%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₇NO₄
MDL Number: MFCD00235883
CAS Number: 150256-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR