Ethyl piperazine-1-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: 1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate

A190805-10G A190805-25G A190805-500G A190805-5G
77111-742EA 6.18 USD
77111-742 77111-744 77111-746 77111-748
Ethyl piperazine-1-carboxylate 96%
Ethyl piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₄N₂O₂
MDL Number: MFCD00005964
CAS Number: 120-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR