3-Methylpyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A189391-10G A189391-1G A189391-250MG A189391-25G A189391-5G
77111-696EA 39.26 USD
77111-696 77111-698 77111-700 77111-702 77111-704
3-Methylpyrazine-2-carboxylic acid 97%
3-Methylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C6H6N2O2
MW: 138.1200 g/mol
MDL Number: MFCD08705765
CAS Number: 41110-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR