VX-745 98%

Supplier: Ambeed
Warning

Synonyms: 5-(2,6-Dichlorophenyl)-2-((2,4-difluorophenyl)thio)-6H-pyrimido[1,6-b]pyridazin-6-one, Neflamapimod

A189058-10MG A189058-1MG A189058-50MG A189058-5MG
77111-686EA 71.09 USD
77111-686 77111-688 77111-690 77111-692
VX-745 98%
VX-745

Precaución: for Research Use Only

Formula: C₁₉H₉Cl₂F₂N₃OS
MW: 436.3 g/mol
MDL Number: MFCD09834070
CAS Number: 209410-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR