2,3,5,6-Tetrachloroisonicotinic acid 95%

Supplier: Ambeed
Danger

A188873-100MG A188873-1G
77111-682EA 13.39 USD
77111-682 77111-684
2,3,5,6-Tetrachloroisonicotinic acid 95%
2,3,5,6-Tetrachloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆HCl₄NO₂
MDL Number: MFCD00092427
CAS Number: 19340-26-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR