3-(4-Iodophenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Iodophenyl)propanoic acid

A187815-10G A187815-1G A187815-250MG A187815-25G A187815-5G A187815-100G
77111-662EA 140.04 USD
77111-662 77111-664 77111-666 77111-668 77111-670 77685-442
3-(4-Iodophenyl)propionic acid 98%
3-(4-Iodophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉IO₂
MW: 276.08 g/mol
MDL Number: MFCD00017597
CAS Number: 1643-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR