3,3-Dimethylbutyraldehyde 95%

Supplier: Ambeed
Danger

Synonyms: 3,3-Dimethylbutanal

A187800-1G A187800-25G A187800-5G
77111-656EA 5.31 USD
77111-656 77111-658 77111-660
3,3-Dimethylbutyraldehyde 95%
3,3-Dimethylbutyraldehyde

Precaución: for Research Use Only

Formula: C₆H₁₂O
MW: 100.16 g/mol
MDL Number: MFCD00042807
CAS Number: 2987-16-8
UN: 1989
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR