Salicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

A187633-1000G A187633-100G A187633-10G A187633-25G A187633-500G
77111-644EA 21.63 USD
77111-644 77111-646 77111-648 77111-650 77111-652
Salicylic acid 98%
Salicylic acid

Precaución: for Research Use Only

Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 336
Flash Pt: 157
MDL Number: MFCD00002439
CAS Number: 69-72-7
Merck Index: 13,08411

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR