8-Fluoro-3,4-dihydro-2H-1,4-benzoxazine 98%

Supplier: Ambeed
Warning

Synonyms: 8-Fluoro-3,4-dihydro-2H-benzo[b][1,4]oxazine

A186600-250MG A186600-1G
77111-602EA 81.69 USD
77111-602 77685-439
8-Fluoro-3,4-dihydro-2H-1,4-benzoxazine 98%
8-Fluoro-3,4-dihydro-2H-1,4-benzoxazine

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.154 g/mol
MDL Number: MFCD08544330
CAS Number: 898832-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR