Ethyl-2-methylacetoacetate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-methyl-3-oxobutanoate
A186472-1000G
A186472-100G
A186472-10G
A186472-25G
A186472-500G
A186472-5G
77111-586EA
95.49
USD
77111-586
77111-588
77111-590
77111-592
77111-594
77111-596
Ethyl-2-methylacetoacetate 95%
Ethyl-2-methylacetoacetate
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol |
MDL Number:
MFCD00009164 CAS Number: 609-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...