Methyl-2-mercaptobenzo[d]oxazole-6-carboxylate 95%

Supplier: Ambeed
Warning

A185951-1G A185951-250MG A185951-25G A185951-5G
77111-578EA 20.6 USD
77111-578 77111-580 77111-582 77111-584
Methyl-2-mercaptobenzo[d]oxazole-6-carboxylate 95%
Methyl-2-mercaptobenzo[d]oxazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₃S
MDL Number: MFCD09261169
CAS Number: 72752-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR