tert-Butylhydrazine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A184637-1000G
A184637-100G
A184637-1G
A184637-25G
A184637-500G
A184637-5G
77111-530EA
50.47
USD
77111-530
77111-532
77111-534
77111-536
77111-538
77111-540
tert-Butylhydrazine hydrochloride 95%
tert-Butylhydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₃ClN₂ |
MDL Number:
MFCD00012947 CAS Number: 7400-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...