Pyrrolidine-3-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A184152-1G A184152-25G A184152-5G
77111-506EA 24.41 USD
77111-506 77111-508 77111-510
Pyrrolidine-3-carboxylic acid hydrochloride 95%
Pyrrolidine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C5H10ClNO2
MW: 151.59 g/mol
MDL Number: MFCD06801272
CAS Number: 953079-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR