Nicotinamidine hydrochloride 95%

Supplier: Ambeed
Warning

A183370-1G A183370-5G
77111-502EA 9.26 USD
77111-502 77567-684
Nicotinamidine hydrochloride 95%
Nicotinamidine hydrochloride

Precaución: For research use only.

Formula: C₆H₈ClN₃
MDL Number: MFCD00054326
CAS Number: 7356-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR