2,4-Diamino-6-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A182627-10G A182627-1G A182627-250MG
77111-458EA 220.67 USD
77111-458 77111-460 77111-462
2,4-Diamino-6-methoxypyrimidine 97%
2,4-Diamino-6-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₈N₄O
MW: 140.1400 g/mol
MDL Number: MFCD02091088
CAS Number: 3270-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR