6-Methyl-2-pyridone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-6-methylpyridine, 6-Methylpyridin-2-ol, 6-Methylpyridin-2(1H)-one

A182009-100G A182009-10G A182009-25G A182009-500G A182009-5G
77111-444EA 88.06 USD
77111-444 77111-446 77111-448 77111-450 77111-452
6-Methyl-2-pyridone 98%
6-Methyl-2-pyridone

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00006279
CAS Number: 3279-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR