GS-9973 99+%

Supplier: Ambeed
Warning

A181632-1MG A181632-50MG A181632-10MG A181632-5MG A181632-100MG
77111-440EA 13.39 USD
77111-440 77111-442 77611-762 77611-764 77685-422
GS-9973 99+%
GS-9973

Precaución: for Research Use Only

Formula: C₂₃H₂₁N₇O
MDL Number: MFCD28099806
CAS Number: 1229208-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR