4-Amino-2,3-dichlorobenzonitrile 95%

Supplier: Ambeed
Warning

A181452-1G A181452-250MG A181452-5G
77111-434EA 43.25 USD
77111-434 77111-436 77111-438
4-Amino-2,3-dichlorobenzonitrile 95%
4-Amino-2,3-dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD20720875
CAS Number: 193090-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR