Potassium 2-(1,3-dioxoisoindolin-2-yl)ethanesulfonate 95%

Supplier: Ambeed
Warning

A181391-1G A181391-250MG A181391-5G
77111-428EA 6.18 USD
77111-428 77111-430 77111-432
Potassium 2-(1,3-dioxoisoindolin-2-yl)ethanesulfonate 95%
Potassium 2-(1,3-dioxoisoindolin-2-yl)ethanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₈KNO₅S
MDL Number: MFCD00023093
CAS Number: 91893-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR