(S)-2-((tert-Butoxycarbonyl)amino)heptanoic acid 95%

Supplier: Ambeed
Danger

A180976-1G A180976-250MG A180976-5G
77111-418EA 55.17 USD
77111-418 77111-420 77111-422
(S)-2-((tert-Butoxycarbonyl)amino)heptanoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)heptanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
MW: 245.32 g/mol
MDL Number: MFCD06659979
CAS Number: 71066-01-8
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR