2-(Naphthalen-1-yl)propan-2-ol 95%

Supplier: Ambeed
Warning

A180643-100MG A180643-1G A180643-250MG
77111-400EA 18.55 USD
77111-400 77111-402 77111-404
2-(Naphthalen-1-yl)propan-2-ol 95%
2-(Naphthalen-1-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₄O
MDL Number: MFCD00085628
CAS Number: 6301-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR