4-(1H-Pyrazol-4-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A179998-100MG A179998-1G A179998-250MG
77111-392EA 60.48 USD
77111-392 77111-394 77111-396
4-(1H-Pyrazol-4-yl)benzoic acid 95%
4-(1H-Pyrazol-4-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD06803014
CAS Number: 1017794-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR