(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%

Supplier: Ambeed
Warning

A179792-100G A179792-10G A179792-25G A179792-500G A179792-5G
77111-374EA 18.55 USD
77111-374 77111-376 77111-378 77111-380 77111-382
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one

Precaución: for Research Use Only

Formula: C₆H₇NO
MDL Number: MFCD00213364
CAS Number: 49805-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR